BIO-Acetoxime 95%

Supplier: Ambeed
Danger

A731227-1MG A731227-5MG
77105-104EA 9 USD
77105-104 77105-106
BIO-Acetoxime 95%
BIO-Acetoxime

Precaución: for Research Use Only

Formula: C₁₈H₁₂BrN₃O₃
MDL Number: MFCD09037533
CAS Number: 667463-85-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR