(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylate, (R)-2-(Aminomethyl)-1-Boc-pyrrolidine, (R)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylic acid tert-Butyl ester

A729696-1G A729696-25G
77105-094EA 12 USD
77105-094 77105-096
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 98%
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03419256
CAS Number: 259537-92-3
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR