2,4-Difluoro-N-(2-nitrophenyl)aniline 95%

Supplier: Ambeed
Warning

A728232-1G A728232-25G A728232-5G
77105-088EA 10 USD
77105-088 77105-090 77105-092
2,4-Difluoro-N-(2-nitrophenyl)aniline 95%
2,4-Difluoro-N-(2-nitrophenyl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₈F₂N₂O₂
MDL Number: MFCD10567165
CAS Number: 500302-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR