1-(6-Bromopyridin-3-yl)cyclopropanecarbonitrile 95%

Supplier: Ambeed
Warning

A728141-100MG A728141-250MG
77105-084EA 73 USD
77105-084 77105-086
1-(6-Bromopyridin-3-yl)cyclopropanecarbonitrile 95%
1-(6-Bromopyridin-3-yl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD20278070
CAS Number: 827628-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR