3,5-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine 95%

Supplier: Ambeed
Warning

A726018-100MG A726018-250MG
77105-064EA 51.99 USD
77105-064 77105-066
3,5-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine 95%
3,5-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₄₁H₂₇N₃
MW: 561.67 g/mol
MDL Number: MFCD20275105
CAS Number: 1013405-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR