4-Chloro-6,8-dimethylquinoline 95%

Supplier: Ambeed
Danger

A725868-100MG A725868-1G A725868-250MG
77105-058EA 28.65 USD
77105-058 77105-060 77105-062
4-Chloro-6,8-dimethylquinoline 95%
4-Chloro-6,8-dimethylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClN
MW: 191.6568 g/mol
MDL Number: MFCD00087699
CAS Number: 196803-72-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR