(4S)-4-Hydroxy-L-isoleucine 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S)-2-Amino-4-hydroxy-3-methylpentanoic acid

A723687-1G A723687-5G
77105-046EA 56 USD
77105-046 77595-140
(4S)-4-Hydroxy-L-isoleucine 97%
(4S)-4-Hydroxy-L-isoleucine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₃
MW: 147.17 g/mol
MDL Number: MFCD07357252
CAS Number: 55399-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR