(5,6-Dimethylpyridin-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A723626-100MG A723626-1G A723626-250MG A723626-5G
77105-040EA 25 USD
77105-040 77105-042 77105-044 77686-721
(5,6-Dimethylpyridin-3-yl)boronic acid ≥95%
(5,6-Dimethylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₂
MDL Number: MFCD11656349
CAS Number: 1370042-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR