3-Fluoro-5-(hydroxymethyl)benzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(hydroxymethyl)phenylboronic acid

A713520-100MG A713520-1G A713520-250MG
77105-010EA 39 USD
77105-010 77105-012 77105-014
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid 97%
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.946 g/mol
MDL Number: MFCD19687060
CAS Number: 1146614-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR