2-Fluoro-4-methyl-5-nitrobenzaldehyde 98%

Supplier: Ambeed
Danger

A713076-1G A713076-250MG A713076-5G
77105-002EA 82.76 USD
77105-002 77105-004 77105-006
2-Fluoro-4-methyl-5-nitrobenzaldehyde 98%
2-Fluoro-4-methyl-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FNO₃
MW: 183.13 g/mol
MDL Number: MFCD28024852
CAS Number: 1804054-69-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR