2,4,6-Triiodo-1,3-benzenediol 95%

Supplier: Ambeed
Warning

A712876-5G A712876-1G A712876-25G
77105-000EA 261 USD
77105-000 77686-701 77900-587
2,4,6-Triiodo-1,3-benzenediol 95%
2,4,6-Triiodo-1,3-benzenediol

Precaución: for Research Use Only

Formula: C₆H₃I₃O₂
MDL Number: MFCD00463616
CAS Number: 19403-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR