tert-Butyl (3-(3-aminopyridin-4-yl)-5-methylcyclohexyl)carbamate 95%

Supplier: Ambeed
Warning

A712381-100MG A712381-250MG
77104-996EA 52 USD
77104-996 77104-998
tert-Butyl (3-(3-aminopyridin-4-yl)-5-methylcyclohexyl)carbamate 95%
tert-Butyl (3-(3-aminopyridin-4-yl)-5-methylcyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₇N₃O₂
MDL Number: MFCD24544027
CAS Number: 1824388-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR