2-Iodo-3-methylpyridine 98%

Supplier: Ambeed
Danger

A712205-25G A712205-1G
77104-994EA 344.8 USD
77104-994 77686-700
2-Iodo-3-methylpyridine 98%
2-Iodo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆IN
MDL Number: MFCD04039350
CAS Number: 22282-58-2
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR