1-(3,5-Difluoropyridin-2-yl)ethanol 95%

Supplier: Ambeed
Warning

A709436-100MG A709436-1G A709436-250MG A709436-5G
77104-982EA 100.79 USD
77104-982 77104-984 77104-986 77104-988
1-(3,5-Difluoropyridin-2-yl)ethanol 95%
1-(3,5-Difluoropyridin-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MW: 159.13 g/mol
MDL Number: MFCD20226879
CAS Number: 1075756-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR