3-Bromo-2,6-difluoronitrobenzene 98%

Supplier: Ambeed
Danger

A708154-1G A708154-25G
77104-968EA 10 USD
77104-968 77104-970
3-Bromo-2,6-difluoronitrobenzene 98%
3-Bromo-2,6-difluoronitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD22689084
CAS Number: 1420800-30-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR