(2,5-Dichloro-4-methoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloro-4-methoxyphenylboronic acid

A704203-100MG A704203-1G A704203-250MG
77104-950EA 47 USD
77104-950 77104-952 77104-954
(2,5-Dichloro-4-methoxyphenyl)boronic acid 98%
(2,5-Dichloro-4-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₃
MW: 220.85 g/mol
MDL Number: MFCD09998846
CAS Number: 1612184-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR