2,5-Diiodobenzene-1,4-diol 95%
Supplier: Ambeed
|
Warning
|
A703520-250MG
A703520-25G
A703520-5G
A703520-1G
77104-940EA
20.16
USD
77104-940
77104-942
77104-944
77573-280
2,5-Diiodobenzene-1,4-diol 95%
2,5-Diiodobenzene-1,4-diol
Precaución: for Research Use Only
Formula:
C₆H₄I₂O₂ MW: 361.90 g/mol |
MDL Number:
MFCD09910095 CAS Number: 13064-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...