2,6-Dibromo-[1,2,4]triazolo[1,5-a]pyridine 95%

Supplier: Ambeed
Warning

A703471-100MG A703471-250MG A703471-1G A703471-5G
77104-936EA 15 USD
77104-936 77104-938 77552-134 77573-278
2,6-Dibromo-[1,2,4]triazolo[1,5-a]pyridine 95%
2,6-Dibromo-[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₃Br₂N₃
MDL Number: MFCD26131653
CAS Number: 1401624-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR