4-Amino-5-chloro-2-methoxy-N-(piperidin-4-ylmethyl)benzamide 97%

Supplier: Ambeed
Warning

A701876-100MG A701876-1G A701876-250MG
77104-922EA 18 USD
77104-922 77104-924 77104-926
4-Amino-5-chloro-2-methoxy-N-(piperidin-4-ylmethyl)benzamide 97%
4-Amino-5-chloro-2-methoxy-N-(piperidin-4-ylmethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₂₀ClN₃O₂
MDL Number: MFCD17212472
CAS Number: 220032-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR