4-Cyclopropyl-benzenemethanol 95%

Supplier: Ambeed
Warning

A698569-250MG A698569-5G A698569-1G
77104-912EA 23 USD
77104-912 77686-682 77900-827
4-Cyclopropyl-benzenemethanol 95%
4-Cyclopropyl-benzenemethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MDL Number: MFCD06802675
CAS Number: 454678-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR