1-Acetyl-3-bromo-4-chloroindole 97%

Supplier: Ambeed
Warning

A697304-250MG A697304-5G
77104-906EA 80.63 USD
77104-906 77104-908
1-Acetyl-3-bromo-4-chloroindole 97%
1-Acetyl-3-bromo-4-chloroindole

Precaución: for Research Use Only

Formula: C₁₀H₇BrClNO
MW: 272.52 g/mol
MDL Number: MFCD22205873
CAS Number: 1375064-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR