4-Benzyl 1-tert-butyl 2-oxopiperazine-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A696932-1G A696932-250MG
77104-902EA 93.36 USD
77104-902 77104-904
4-Benzyl 1-tert-butyl 2-oxopiperazine-1,4-dicarboxylate 95%
4-Benzyl 1-tert-butyl 2-oxopiperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₅
MW: 334.36 g/mol
MDL Number: MFCD28054177
CAS Number: 1228675-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR