4-Isopropylsalicylaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-isopropylbenzaldehyde

A696635-1G A696635-250MG
77104-898EA 71.09 USD
77104-898 77104-900
4-Isopropylsalicylaldehyde 97%
4-Isopropylsalicylaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
MDL Number: MFCD16997144
CAS Number: 536-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR