1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole, 1-Methyl-1h-indazol-3-ylboronic acid pinacol ester

A695557-100MG A695557-1G A695557-250MG
77104-892EA 66 USD
77104-892 77104-894 77104-896
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 98%
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD13182067
CAS Number: 1627722-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR