Ethyl-3,5-difluorobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,5-difluorobenzoate

A694793-100G A694793-25G A694793-5G
77104-886EA 162.32 USD
77104-886 77104-888 77104-890
Ethyl-3,5-difluorobenzoate 95%
Ethyl-3,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD03093801
CAS Number: 350-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR