Benzyl (4-bromo-2-chlorophenyl)carbamate 98%

Supplier: Ambeed
Warning

A694544-1G A694544-250MG A694544-5G
77104-880EA 79 USD
77104-880 77104-882 77104-884
Benzyl (4-bromo-2-chlorophenyl)carbamate 98%
Benzyl (4-bromo-2-chlorophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrClNO₂
MW: 340.6 g/mol
MDL Number: MFCD18088428
CAS Number: 1259064-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR