m-Tolylacetonitrile 98%

Supplier: Ambeed
Danger

A693278-10G A693278-25G A693278-5G
77104-870EA 11 USD
77104-870 77104-872 77104-874
m-Tolylacetonitrile 98%
m-Tolylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₉N
MDL Number: MFCD00001914
CAS Number: 2947-60-6
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR