Fmoc-L-allo-isoleucine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-allo-Ile-OH, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylpentanoic acid
A692282-100MG
A692282-1G
A692282-250MG
A692282-5G
77104-858EA
61
USD
77104-858
77104-860
77104-862
77901-711
Fmoc-L-allo-isoleucine 98%
Fmoc-L-allo-isoleucine
Precaución: for Research Use Only
Formula:
C₂₁H₂₃NO₄ MW: 353.418 g/mol |
MDL Number:
MFCD01631976 CAS Number: 251316-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...