(R)-tert-Butyl (1-benzylpiperidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A691795-1G A691795-250MG
77104-856EA 47 USD
77104-856 77901-726
(R)-tert-Butyl (1-benzylpiperidin-3-yl)carbamate 97%
(R)-tert-Butyl (1-benzylpiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₆N₂O₂
MDL Number: MFCD07369848
CAS Number: 454713-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR