2-Bromo-5-iodonitrobenzene 98%

Supplier: Ambeed
Warning

A691323-1G A691323-250MG A691323-5G
77104-852EA 15 USD
77104-852 77104-854 77901-342
2-Bromo-5-iodonitrobenzene 98%
2-Bromo-5-iodonitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrINO₂
MDL Number: MFCD12547765
CAS Number: 713512-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR