α-Bromo-2,6-dichlorotoluene 98%

Supplier: Ambeed
Danger

A688343-25G A688343-5G A688343-10G
77104-842EA 22 USD
77104-842 77104-844 77686-668
α-Bromo-2,6-dichlorotoluene 98%
α-Bromo-2,6-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MDL Number: MFCD00000577
CAS Number: 20443-98-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR