1-Methyl-5-indoleboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylindole-5-boronic acid, N-Methylindole-5-boronic acid

A688259-1G A688259-250MG A688259-5G
77104-836EA 47.75 USD
77104-836 77104-838 77104-840
1-Methyl-5-indoleboronic acid 98%
1-Methyl-5-indoleboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₂
MW: 174.9900 g/mol
MDL Number: MFCD03412070
CAS Number: 192182-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR