4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethynyl)benzoate

A682866-100MG A682866-1G A682866-250MG
77104-808EA 30.77 USD
77104-808 77104-810 77104-812
4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester 95+%
4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₂H₂₃BO₄
MW: 362.23 g/mol
MDL Number: MFCD16294527
CAS Number: 1448637-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR