tert-Butyl 2-methyl-1H-pyrrole-1-carboxylate 95%

Supplier: Ambeed
Warning

A682554-250MG A682554-1G
77104-806EA 45.62 USD
77104-806 77614-108
tert-Butyl 2-methyl-1H-pyrrole-1-carboxylate 95%
tert-Butyl 2-methyl-1H-pyrrole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MW: 181.23 g/mol
MDL Number: MFCD21362405
CAS Number: 72590-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR