2,4-Diiodoaniline 95%

Supplier: Ambeed
Warning

A681784-1G A681784-250MG A681784-5G
77104-800EA 21.99 USD
77104-800 77104-802 77104-804
2,4-Diiodoaniline 95%
2,4-Diiodoaniline

Precaución: for Research Use Only

Formula: C₆H₅I₂N
MDL Number: MFCD01851096
CAS Number: 533-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR