4-(2-Chloro-9H-purin-6-yl)morpholine 95%

Supplier: Ambeed
Warning

A679575-1G A679575-250MG
77104-796EA 91.24 USD
77104-796 77104-798
4-(2-Chloro-9H-purin-6-yl)morpholine 95%
4-(2-Chloro-9H-purin-6-yl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₀ClN₅O
MW: 239.66 g/mol
MDL Number: MFCD17430374
CAS Number: 4010-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR