(4-Fluoro-3-methoxyphenyl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A679569-1G A679569-250MG
77104-792EA 28 USD
77104-792 77104-794
(4-Fluoro-3-methoxyphenyl)hydrazine hydrochloride 95%
(4-Fluoro-3-methoxyphenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀ClFN₂O
MDL Number: MFCD14583037
CAS Number: 327021-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR