(2R,3R)-2-(2,5-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol 95%

Supplier: Ambeed
Warning

A679447-100MG A679447-250MG A679447-25G A679447-5G
77104-784EA 11.67 USD
77104-784 77104-786 77104-788 77104-790
(2R,3R)-2-(2,5-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol 95%
(2R,3R)-2-(2,5-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂N₃O₂
MW: 269.24 g/mol
MDL Number: MFCD28404614
CAS Number: 241479-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR