4-Methoxy-3-(methoxycarbonyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

A678135-100MG A678135-1G A678135-250MG A678135-5G
77104-774EA 16 USD
77104-774 77104-776 77104-778 77686-647
4-Methoxy-3-(methoxycarbonyl)phenylboronic acid 95%
4-Methoxy-3-(methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MDL Number: MFCD23380361
CAS Number: 1071958-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR