tert-Butyl 4-(pyrimidin-2-yl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A677107-100G A677107-1G A677107-25G A677107-5G
77104-764EA 1221.1 USD
77104-764 77104-766 77104-768 77104-770
tert-Butyl 4-(pyrimidin-2-yl)piperazine-1-carboxylate 97%
tert-Butyl 4-(pyrimidin-2-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₄O₂
MW: 264.32 g/mol
MDL Number: MFCD12068359
CAS Number: 780705-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR