6-Isopropoxynaphthalene-2-boronic acid 98%

Supplier: Ambeed
Warning

A675404-1G A675404-250MG A675404-5G
77104-748EA 108 USD
77104-748 77104-750 77104-752
6-Isopropoxynaphthalene-2-boronic acid 98%
6-Isopropoxynaphthalene-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅BO₃
MDL Number: MFCD16260719
CAS Number: 1334221-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR