2-Chloro-4-iodoanisole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-iodo-1-methoxybenzene

A672947-5G A672947-1G A672947-250MG
77104-732EA 125 USD
77104-732 77686-637 77900-921
2-Chloro-4-iodoanisole 95%
2-Chloro-4-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆ClIO
MDL Number: MFCD15527196
CAS Number: 75676-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR