4-Iodo-3-nitrophenol 98%

Supplier: Ambeed
Danger

A672185-1G A672185-250MG A672185-5G
77104-724EA 25 USD
77104-724 77104-726 77104-728
4-Iodo-3-nitrophenol 98%
4-Iodo-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄INO₃
MDL Number: MFCD01320680
CAS Number: 113305-56-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR