1-Iodo-2-methoxy-ethane 95%

Supplier: Ambeed
Danger

A671660-100MG A671660-1G A671660-250MG A671660-5G
77104-716EA 5 USD
77104-716 77104-718 77104-720 77104-722
1-Iodo-2-methoxy-ethane 95%
1-Iodo-2-methoxy-ethane

Precaución: for Research Use Only

Formula: C₃H₇IO
MDL Number: MFCD07644549
CAS Number: 4296-15-5
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR