4-(Methylsulfonyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylsulphonyl)benzyl Alcohol

A670469-10G A670469-1G A670469-25G A670469-5G
77104-704EA 37.14 USD
77104-704 77104-706 77104-708 77104-710
4-(Methylsulfonyl)benzyl alcohol 97%
4-(Methylsulfonyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
MDL Number: MFCD03425695
CAS Number: 22821-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR