5-Amino-2,4-dichlorophenol 98%

Supplier: Ambeed
Danger

A668953-1G A668953-250MG A668953-5G
77104-696EA 60.48 USD
77104-696 77104-698 77104-700
5-Amino-2,4-dichlorophenol 98%
5-Amino-2,4-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.012 g/mol
MDL Number: MFCD00186187
CAS Number: 39489-79-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR