2-Methylimidazo[1,2-a]pyridine-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A668679-100MG A668679-250MG A668679-1G
77104-684EA 96.55 USD
77104-684 77104-686 77686-635
2-Methylimidazo[1,2-a]pyridine-6-carboxylic acid 95%
2-Methylimidazo[1,2-a]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD09258863
CAS Number: 129912-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR