(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-iodophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A667477-1G A667477-250MG
77104-670EA 82.76 USD
77104-670 77104-672 77104-674
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-iodophenyl)propanoic acid 98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-iodophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₀INO₄
MDL Number: MFCD06796745
CAS Number: 478183-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR