2,3',5'-Trifluoro-[1,1'-biphenyl]-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A667037-100MG A667037-1G A667037-250MG
77104-664EA 61.54 USD
77104-664 77104-666 77104-668
2,3',5'-Trifluoro-[1,1'-biphenyl]-4-carbaldehyde 97%
2,3',5'-Trifluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇F₃O
MW: 236.18 g/mol
MDL Number: MFCD25460352
CAS Number: 1318975-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR