1-Bromo-2,3,4-trifluorobenzene 98%

Supplier: Ambeed
Warning

A666158-1000G A666158-100G A666158-10G A666158-25G A666158-500G A666158-5G
77104-650EA 290.69 USD
77104-650 77104-652 77104-654 77104-656 77104-658 77104-660
1-Bromo-2,3,4-trifluorobenzene 98%
1-Bromo-2,3,4-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
MDL Number: MFCD00013248
CAS Number: 176317-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR